missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(1-methoxy-2-methyl-2-propoxy)bismuth, 99.99% (metals basis)

Manufacturer:  Alfa Aesar™ 4548814

 View more versions of this product

Catalog No. AA4548814



Description

Description

  • Packaged under Argon
Specifications

Specifications

Tris (1-methoxy-2-methyl-2-propoxy)bismuth
Yellow
(metals basis)
MFCD11113030
Crystalline
25g
70°C
C15H33O6Bi
518.4
99.99%

Chemical Identifiers

C15H33O6Bi
MFCD11113030
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 :

  • Packaged under Argon

SDS Documents