missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine, TCI America™

Manufacturer:  TCI America T29935G

 View more versions of this product

Catalog No. T29935G



Specifications

Specifications

Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine
138°C
510758-28-8
MFCD09265124
WKGZJBVXZWCZQC-UHFFFAOYSA-N
1-(1-benzyltriazol-4-yl)-N,N-bis[(1-benzyltriazol-4-yl)methyl]methanamine
11203363
Crystal-Powder at 20°C
Undesignated
5g
C30H30N10
tris 1-benzyl-1h-1,2,3-triazol-4-yl methyl amine, tris 1-benzyl-1h-1,2,3-triazol-4-yl methyl amine, 1,2,4-trimethyl-5-benzenesulfonic acid, tris 1-benzyl-1h-1,2,3-triazole-4-ylmethyl amine, tris 1-benzyl-1h-1,2,3-triazol-4-yl methyl amine tbta, tris 1-benzyl-1h-1,2,3-triazol-4-yl methyl amine
C1=CC=C(C=C1)CN2C=C(N=N2)CN(CC3=CN(N=N3)CC4=CC=CC=C4)CC5=CN(N=N5)CC6=CC=CC=C6
530.64
530.64
≥97.0% (HPLC,T)

Chemical Identifiers

510758-28-8
530.64
WKGZJBVXZWCZQC-UHFFFAOYSA-N
11203363
C1=CC=C(C=C1)CN2C=C(N=N2)CN(CC3=CN(N=N3)CC4=CC=CC=C4)CC5=CN(N=N5)CC6=CC=CC=C6
C30H30N10
MFCD09265124
tris 1-benzyl-1h-1,2,3-triazol-4-yl methyl amine, tris 1-benzyl-1h-1,2,3-triazol-4-yl methyl amine, 1,2,4-trimethyl-5-benzenesulfonic acid, tris 1-benzyl-1h-1,2,3-triazole-4-ylmethyl amine, tris 1-benzyl-1h-1,2,3-triazol-4-yl methyl amine tbta, tris 1-benzyl-1h-1,2,3-triazol-4-yl methyl amine
1-(1-benzyltriazol-4-yl)-N,N-bis[(1-benzyltriazol-4-yl)methyl]methanamine
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents