Please login to your online account to display your discounted pricing

(Tris)-1,10-Phenanthroline Iron (II) Sulfate Solution (0.025M/Certified), Fisher Chemical

$99.75 - $207.25

Chemical Identifiers

Chemical Name or Material (Tris)-1, 10-Phenanthroline Iron (II) Sulfate Solution
Name Note 0.025M
CAS 7732-18-5
CAS 14634-91-4
CAS 7782-63-0
Purity Grade Certified Grade
Molecular Formula C36H24FeN6O4S
Linear Formula (C12H8N2)3FeSO4
MDL Number MFCD00036428
Synonym Ferroin
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

P69-100

  SDS
fisher scientific company
P69100
Glass Bottle 100mL Each for $99.75
Case of 6 Each for $598.50

P69-500

  SDS
fisher scientific company
P69500
Glass Bottle 500mL Each for $207.25
Case of 6 Each for $1,243.50
Description & Specifications

Specifications

Purity Grade Certified Grade
Quantity 100mL
Physical Form Liquid
Color Dark Red
Melting Point 0°C
Vapor Pressure 14mmHg
Alpha Vector Phenanthroline Iron (II) Sulfate Solution
Density 1.0g/cm³
Packaging Glass Bottle
CAS 14634-91-4
MDL Number MFCD00036428
Identification Pass Test
Synonym Ferroin
CAS 7782-63-0
Suitability Pass Test (redox indicator)
ChemAlert Storage Symbol Gray
CAS 7732-18-5
Chemical Name or Material (Tris)-1, 10-Phenanthroline Iron (II) Sulfate Solution
Linear Formula (C12H8N2)3FeSO4
Molecular Formula C36H24FeN6O4S
Name Note 0.025M

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

Safety and Handling