Please login to your online account to display your discounted pricing

Tris(1,10-phenanthroline)iron(II) sulfate, ACROS Organics™


Quantity 100mL
Packaging Glass bottle

Manufacturer: fisher scientific acros  422251000

 View more versions of this product

Catalog No. AC422251000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 692.53
Quantity 100mL
Color Dark red
Density 1.0060g/mL
Chemical Name or Material Tris(1, 10-phenanthroline)iron(II) sulfate
Name Note 0.025M Aqueous Solution
Molecular Formula C36H24FeN6O4S
Packaging Glass bottle
MDL Number MFCD00036428
CAS 14634-91-4
Suitability (for use as a indicator for iron) passes
Synonym Ferroin indicator solution

Specifications

Chemical Name or Material Tris(1, 10-phenanthroline)iron(II) sulfate
Name Note 0.025M Aqueous Solution
CAS 14634-91-4
Molecular Formula C36H24FeN6O4S
Formula Weight 692.53
MDL Number MFCD00036428
Synonym Ferroin indicator solution