Please login to your online account to display your discounted pricing

Alfa Aesar™ TRIS, 1.0M buffer soln., pH 7.5

$71.88 - $71.88

Chemical Identifiers

Chemical Name or Material TRIS buffer soln.
CAS 77-86-1
MDL Number MFCD00004679
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ60636K2

  SDS
alfa aesar™
J60636K2
1L Each for $71.88
Description & Specifications

Specifications

Chemical Name or Material TRIS buffer soln.
CAS 77-86-1
MDL Number MFCD00004679
Physical Form Liquid
TSCA TSCA
Odor Odorless
Concentration 1.0M buffer soln.
Quantity 1L
pH 7.5

Applications
TRIS, 1.0M buffer soln., pH 7.5 is used as buffers for stabilizing and storing nucleic acids and proteins.

Solubility
Soluble in water and organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Hygroscopic. Store under inert gas. Keep away fom strong oxidizing agents.