Please login to your online account to display your discounted pricing

Alfa Aesar™ TRIS, 1.0M buffer soln., pH 6.8

$71.88 - $71.88

Chemical Identifiers

Chemical Name or Material TRIS buffer soln.
CAS 77-86-1
MDL Number MFCD00004679
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ63831K2

  SDS
alfa aesar™
J63831K2
1L Each for $71.88
Description & Specifications

Specifications

Chemical Name or Material TRIS buffer soln.
CAS 77-86-1
Physical Form Liquid
TSCA TSCA
pH 6.8
Odor Odorless
Quantity 1L
Concentration 1.0M buffer soln.
MDL Number MFCD00004679

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.