missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ TRIS, 0.5M buffer soln., pH 8.5

$93.30 - $133.24

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
MDL Number MFCD00004679
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ62131AP View Documents Alfa Aesar™
J62131AP
500mL Each for $93.30
 
AAJ62131K2 View Documents Alfa Aesar™
J62131K2
1L Each for $133.24
 
Description

Description

Applications
TRIS, 1.0M buffer soln., pH 7.5 is used as buffers for stabilizing and storing nucleic acids and proteins.

Solubility
Soluble in water and organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Hygroscopic. Store under inert gas. Keep away fom strong oxidizing agents.
Specifications

Chemical Identifiers

77-86-1
121.136
LENZDBCJOHFCAS-UHFFFAOYSA-N
CHEBI:9754
C(C(CO)(CO)N)O
C4H11NO3
MFCD00004679
6503
2-amino-2-(hydroxymethyl)propane-1,3-diol

Specifications

77-86-1
MFCD00004679
LENZDBCJOHFCAS-UHFFFAOYSA-N
2-amino-2-(hydroxymethyl)propane-1,3-diol
6503
Liquid
500mL
C4H11NO3
0.5M buffer soln.
C(C(CO)(CO)N)O
121.136
CHEBI:9754
8.5
TRIS buffer soln.
Safety and Handling

Safety and Handling

TSCA : TSCA