missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ TRIS, 0.5M buffer soln., pH 8.4

$59.51 - $84.96

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
MDL Number MFCD00004679
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61537AP View Documents Alfa Aesar™
J61537AP
500mL Each for $59.51
 
AAJ61537K2 View Documents Alfa Aesar™
J61537K2
1L Each for $84.96
 
Specifications

Chemical Identifiers

77-86-1
121.136
LENZDBCJOHFCAS-UHFFFAOYSA-N
CHEBI:9754
C(C(CO)(CO)N)O
C4H11NO3
MFCD00004679
6503
2-amino-2-(hydroxymethyl)propane-1,3-diol

Specifications

77-86-1
MFCD00004679
LENZDBCJOHFCAS-UHFFFAOYSA-N
2-amino-2-(hydroxymethyl)propane-1,3-diol
6503
Liquid
500mL
C4H11NO3
0.5M buffer soln.
C(C(CO)(CO)N)O
121.136
CHEBI:9754
8.4
TRIS buffer soln.
Safety and Handling

Safety and Handling

TSCA : TSCA