missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

TRIS, 0.5M buffer soln., pH 7.2, Thermo Scientific™

$149.81 - $149.81

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
MDL Number MFCD00004679
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61009K2
View Documents Promotion Details
Thermo Scientific Chemicals
J61009K2
1 L
Each for $149.81
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

77-86-1
121.136
LENZDBCJOHFCAS-UHFFFAOYSA-N
CHEBI:9754
C(C(CO)(CO)N)O
C4H11NO3
MFCD00004679
6503
2-amino-2-(hydroxymethyl)propane-1,3-diol

Specifications

77-86-1
7.2
1 L
MFCD00004679
C(C(CO)(CO)N)O
121.136
CHEBI:9754
0.5 M
Liquid
C4H11NO3
LENZDBCJOHFCAS-UHFFFAOYSA-N
2-amino-2-(hydroxymethyl)propane-1,3-diol
6503
TRIS buffer soln.
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'