missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Triruthenium dodecacarbonyl, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 224000050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Triruthenium dodecacarbonyl | |
150.0°C | |
98.5% min. | |
99% | |
C12O12Ru3 | |
MFCD00011209 | |
Ruthenium carbonyl | |
NQZFAUXPNWSLBI-UHFFFAOYSA-N | |
cyclotriruthenium; dodecakis(methanidylidyneoxidanium) | |
639.33 | |
Crystals |
15243-33-1 | |
Orange | |
Authentic | |
Glass bottle | |
Ru3(CO)12 | |
5 g | |
Solubility in water: insoluble. | |
[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Ru]1[Ru][Ru]1 | |
639.33 | |
99% |
Chemical Identifiers
15243-33-1 | |
639.33 | |
NQZFAUXPNWSLBI-UHFFFAOYSA-N | |
cyclotriruthenium; dodecakis(methanidylidyneoxidanium) |
C12O12Ru3 | |
MFCD00011209 | |
Ruthenium carbonyl | |
[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Ru]1[Ru][Ru]1 |
Safety and Handling
EINECSNumber : 239-287-4
RUO – Research Use Only