missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tripropylamine, 98%, ACROS Organics™

Chemical Identifiers

Assay Percent Range 98%
Molecular Formula C9H21N
Linear Formula (CH3CH2CH2)3N
Formula Weight 143.27
MDL Number MFCD00009363
Chemical Name or Material Tripropylamine
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC162690025 SDS Thermo Fisher Scientific
AC162690025
Glass bottle 2.5L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC162690010 SDS Thermo Fisher Scientific
162690010
Glass bottle 1L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

98%
(CH3CH2CH2)3N
MFCD00009363
≥97.5%
C9H21N
143.27
Tripropylamine

Specifications

143.27
2.5L
0.7600g/mL
C9H21N
Glass bottle
0.2% max.
Authentic
102-69-2
Tripropylamine
Liquid
Clear colorless
04, II, 623
MFCD00009363
1.4155 to 1.4175 (20°C, 589nm)
(CH3CH2CH2)3N
dipropylamine: < 0.2%
98%
≥97.5%

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Causes serious eye damage.
Harmful to aquatic life with long lasting effects.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger