missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tripropyl Orthoformate 98.0+%, TCI America™

Manufacturer:  TCI America O025925ML

 View more versions of this product

Catalog No. O025925ML



Specifications

Specifications

Tripropyl Orthoformate
Yellow
621-76-1
MFCD00015214
RWNXXQFJBALKAX-UHFFFAOYSA-N
1-(dipropoxymethoxy)propane
69311
Clear Liquid at 20°C
198°C
25mL
C10H22O3
1-dipropoxymethoxy propane
CCCOC(OCCC)OCCC
190.283
190.28
≥98.0% (GC)

Chemical Identifiers

621-76-1
190.283
RWNXXQFJBALKAX-UHFFFAOYSA-N
69311
CCCOC(OCCC)OCCC
C10H22O3
MFCD00015214
1-dipropoxymethoxy propane
1-(dipropoxymethoxy)propane
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3932

TSCA : Yes

Documents