missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

β-Nicotinamide adenine dinucleotide phosphate sodium salt hydrate, 97+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC227571000

 View more versions of this product

Catalog No. AC227571000



Description

Description

Specifications

Specifications

0.80 to 0.86 (A250/A260,pH 7.5),0.18 to 0.24 (A280/A260,pH 7.5)
Authentic
6% Max. (K.F.)
175.0°C to 178.0°C
698999-85-8
C21H27N7NaO17P3·xH2O
14,6348
Solubility in water: soluble. Other solubilities: soluble in methanol,less soluble in ethanol,practically insoluble in ether and ethyl acetate
C1=CC(=C[N+](=C1)C2C(C(C(O2)COP(=O)([O-])OP(=O)(O)OCC3C(C(C(O3)N4C=NC5=C4N=CN=C5N)OP(=O)(O)O)O)O)O)C(=O)N.[Na]
765.39
765.39
97+%
β-Nicotinamide adenine dinucleotide phosphate sodium salt hydrate
Glass bottle
White to Yellow
100mg
determination,based on dry weight)
MFCD00150480
beta-nicotinamide adenine dinucleotide phosphate sodium, nadp sodium salt
ZUXQIVQXILFYQI-QYZPTAICSA-N
[[(2R,3R,4R,5R)-5-(6-aminopurin-9-yl)-3-hydroxy-4-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate;sodium
131845262
Powder

Chemical Identifiers

698999-85-8
765.39
ZUXQIVQXILFYQI-QYZPTAICSA-N
131845262
C1=CC(=C[N+](=C1)C2C(C(C(O2)COP(=O)([O-])OP(=O)(O)OCC3C(C(C(O3)N4C=NC5=C4N=CN=C5N)OP(=O)(O)O)O)O)O)C(=O)N.[Na]
C21H27N7NaO17P3·xH2O
MFCD00150480
beta-nicotinamide adenine dinucleotide phosphate sodium, nadp sodium salt
[[(2R,3R,4R,5R)-5-(6-aminopurin-9-yl)-3-hydroxy-4-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate;sodium
Safety and Handling

Safety and Handling

EINECSNumber : 214-664-6

SDS Documents