missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
β-Nicotinamide adenine dinucleotide phosphate sodium salt hydrate, 97+%, ACROS Organics™
Manufacturer: Fisher Scientific Acros 227575000
Specifications
0.80 to 0.86 (A250/A260,pH 7.5),0.18 to 0.24 (A280/A260,pH 7.5) | |
Authentic | |
6% Max. (K.F.) | |
175.0°C to 178.0°C | |
698999-85-8 | |
C21H27N7NaO17P3·xH2O | |
14,6348 | |
Solubility in water: soluble. Other solubilities: soluble in methanol,less soluble in ethanol,practically insoluble in ether and ethyl acetate | |
C1=CC(=C[N+](=C1)C2C(C(C(O2)COP(=O)([O-])OP(=O)(O)OCC3C(C(C(O3)N4C=NC5=C4N=CN=C5N)OP(=O)(O)O)O)O)O)C(=O)N.[Na] | |
765.39 | |
765.39 | |
97+% |
β-Nicotinamide adenine dinucleotide phosphate sodium salt hydrate | |
Glass bottle | |
White to Yellow | |
500mg | |
determination,based on dry weight) | |
MFCD00150480 | |
beta-nicotinamide adenine dinucleotide phosphate sodium, nadp sodium salt | |
ZUXQIVQXILFYQI-QYZPTAICSA-N | |
[[(2R,3R,4R,5R)-5-(6-aminopurin-9-yl)-3-hydroxy-4-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate;sodium | |
131845262 | |
Powder |
Chemical Identifiers
698999-85-8 | |
765.39 | |
ZUXQIVQXILFYQI-QYZPTAICSA-N | |
131845262 | |
C1=CC(=C[N+](=C1)C2C(C(C(O2)COP(=O)([O-])OP(=O)(O)OCC3C(C(C(O3)N4C=NC5=C4N=CN=C5N)OP(=O)(O)O)O)O)O)C(=O)N.[Na] |
C21H27N7NaO17P3·xH2O | |
MFCD00150480 | |
beta-nicotinamide adenine dinucleotide phosphate sodium, nadp sodium salt | |
[[(2R,3R,4R,5R)-5-(6-aminopurin-9-yl)-3-hydroxy-4-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate;sodium |
Safety and Handling
EINECSNumber : 214-664-6