Please login to your online account to display your discounted pricing

Triphosphopyridine nucleotide, disodium salt, 97+%, ACROS Organics™


Quantity 500mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  36056-5000

 View more versions of this product

Catalog No. AC360565000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.0% (enzymatic, on dry subst.)
Physical Form Powder
Formula Weight 787.4
Quantity 500mg
Color Yellow
EINECS Number 246-129-8
Solubility Solubility in water: >50g/L water.
Water 6% Max.
Chemical Name or Material Triphosphopyridine nucleotide, disodium salt
Infrared Spectrum Authentic
Assay Percent Range 97+%
Molecular Formula C21H26N7Na2O17P3
Packaging Glass bottle
MDL Number MFCD00065390
CAS 24292-60-2

Specifications

Chemical Name or Material Triphosphopyridine nucleotide, disodium salt
CAS 24292-60-2
Assay ≥97.0% (enzymatic, on dry subst.)
Assay Percent Range 97+%
Molecular Formula C21H26N7Na2O17P3
Formula Weight 787.4
MDL Number MFCD00065390