Learn More
Triphosgene, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 258950500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent, C-X bond formation reagentSpecifications
Triphosgene | |
77.0°C to 81.0°C | |
1.7800g/mL | |
Authentic | |
Glass bottle | |
C(Cl3)OC(O)OC(Cl)3 | |
17,127 | |
15,9921 | |
Solubility in water: practically insoluble, may form phosgene. Other solubilities: 588g/L ch2cl2 (20°C), soluble in alcohol, benzene, tetrachloromethane, and ether | |
C(=O)(OC(Cl)(Cl)Cl)OC(Cl)(Cl)Cl | |
296.75 | |
296.75 | |
Crystalline Powder, Crystals or Chunks |
32315-10-9 | |
White | |
203.0°C to 206.0°C | |
98.5% min | |
C3Cl6O3 | |
50 g | |
1.78 | |
triphosgene, bis trichloromethyl carbonate, methanol, trichloro-, carbonate 2:1, ditrichloromethyl carbonate, triphosgene bis-trichloromethyl carbonate, methanol, 1,1,1-trichloro-, 1,1'-carbonate, triphosgen, tri phosgene, tri-phosgene | |
UCPYLLCMEDAXFR-UHFFFAOYSA-N | |
bis(trichloromethyl) carbonate | |
94429 | |
99% |
Chemical Identifiers
32315-10-9 | |
296.75 | |
triphosgene, bis trichloromethyl carbonate, methanol, trichloro-, carbonate 2:1, ditrichloromethyl carbonate, triphosgene bis-trichloromethyl carbonate, methanol, 1,1,1-trichloro-, 1,1'-carbonate, triphosgen, tri phosgene, tri-phosgene | |
bis(trichloromethyl) carbonate |
C3Cl6O3 | |
UCPYLLCMEDAXFR-UHFFFAOYSA-N | |
94429 | |
C(=O)(OC(Cl)(Cl)Cl)OC(Cl)(Cl)Cl |
Safety and Handling
GHS H Statement
Fatal if inhaled.
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 250-986-3
RUO – Research Use Only