missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triphosgene, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 258950100

 View more versions of this product

Catalog No. AC258950100



Description

Description

Coupling reagent, C-X bond formation reagent
Specifications

Specifications

Triphosgene
Authentic
1.78
White
10g
98.5% min
C(Cl3)OC(O)OC(Cl)3
15,9921
Solubility in water: practically insoluble, may form phosgene. Other solubilities: 588g/L ch2cl2 (20°C), soluble in alcohol, benzene, tetrachloromethane, and ether
C(=O)(OC(Cl)(Cl)Cl)OC(Cl)(Cl)Cl
296.75
296.75
99%
1.7800g/mL
Glass bottle
203.0°C to 206.0°C
77.0°C to 81.0°C
32315-10-9
C3Cl6O3
17,127
bis trichloromethyl carbonate
UCPYLLCMEDAXFR-UHFFFAOYSA-N
bis(trichloromethyl) carbonate
94429
Crystalline Powder, Crystals and/or Chunks

Chemical Identifiers

32315-10-9
296.75
bis trichloromethyl carbonate
bis(trichloromethyl) carbonate
C3Cl6O3
UCPYLLCMEDAXFR-UHFFFAOYSA-N
94429
C(=O)(OC(Cl)(Cl)Cl)OC(Cl)(Cl)Cl
Safety and Handling

Safety and Handling

GHS H Statement
Fatal if inhaled.
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 250-986-3

SDS Documents