missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triphenylvinylsilane, TCI America™

Manufacturer:  TCI America T16955G

 View more versions of this product

Catalog No. T16955G



Specifications

Specifications

Triphenylvinylsilane
White-Yellow
5g
C20H18Si
triphenylvinylsilane, silane, ethenyltriphenyl, triphenyl vinyl silane, silane, triphenylvinyl, vinyltriphenylsilane, benzene, 1,1',1-ethenylsilylidyne tris, ethenyltriphenylsilan, vinyltriphenyl silane, vinyl triphenyl silane, ethenyl triphenyl silane
C=C[Si](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3
286.449
286.45
≥93.0% (GC)
190°C
69°C
18666-68-7
MFCD00051542
OVOIHGSHJGMSMZ-UHFFFAOYSA-N
ethenyl(triphenyl)silane
87747
Crystal-Powder at 20°C

Chemical Identifiers

18666-68-7
286.449
OVOIHGSHJGMSMZ-UHFFFAOYSA-N
87747
C=C[Si](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3
C20H18Si
MFCD00051542
triphenylvinylsilane, silane, ethenyltriphenyl, triphenyl vinyl silane, silane, triphenylvinyl, vinyltriphenylsilane, benzene, 1,1',1-ethenylsilylidyne tris, ethenyltriphenylsilan, vinyltriphenyl silane, vinyl triphenyl silane, ethenyl triphenyl silane
ethenyl(triphenyl)silane
Safety and Handling

Safety and Handling

TSCA : Yes

Documents