missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Triphenyltin hydroxide

Manufacturer:  Alfa Aesar™ 7117714

 View more versions of this product

Catalog No. AA7117714



Specifications

Specifications

Triphenyltin hydroxide
76-87-9
White
(C6H5)3SnOH
UN3146
triphenyltin hydroxide, fentin hydroxide, triphenylstannanol, vancide ks, hydroxytriphenyltin, hydroxytriphenylstannane, erithane, fenolovo, tenhide, stannane, hydroxytriphenyl
NRHFWOJROOQKBK-UHFFFAOYSA-N
triphenyltin;hydrate
6327657
Powder
122°C to 124°C
25g
C18H17OSn
MFCD00013928
14,9745
Slightly soluble in alcohol, toluene
C1=CC=C(C=C1)[Sn](C2=CC=CC=C2)C3=CC=CC=C3.O
368.043
367.01

Chemical Identifiers

76-87-9
368.043
NRHFWOJROOQKBK-UHFFFAOYSA-N
6327657
C1=CC=C(C=C1)[Sn](C2=CC=CC=C2)C3=CC=CC=C3.O
C18H17OSn
MFCD00013928
triphenyltin hydroxide, fentin hydroxide, triphenylstannanol, vancide ks, hydroxytriphenyltin, hydroxytriphenylstannane, erithane, fenolovo, tenhide, stannane, hydroxytriphenyl
triphenyltin;hydrate
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H330-H372-H351-H361-H318-H315-H335
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
Suspected of damaging fertility or the unborn child.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
P260-P301+P310-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.

RTECSNumber : WH8575000

TSCA : TSCA

SDS Documents