missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Triphenyltin hydride, 97%

Manufacturer:  Alfa Aesar™ L0016314

 View more versions of this product

Catalog No. AAL0016314



Description

Description

Applications
Triphenyltin hydride is used as a source of ″H·″ to generate radicals or cleave carbon-oxygen bonds. And it is also used in anti-fouling paints.

Solubility
Insoluble in water.

Notes
Air & Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

Triphenyltin hydride
>110°C (230°F)
Air and moisture sensitive
26°C to 28°C
892-20-6
(C6H5)3SnH
UN3146
Triphenylstannane
351.02
1.374
1.637
162°C to 168°C (0.5mmHg)
25g
C18H16Sn
MFCD00003004
3544353
Insoluble in water.
97%

Chemical Identifiers

892-20-6
C18H16Sn
MFCD00003004
Triphenylstannane
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P501c

H301+H311+H331

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.

EINECSNumber : 212-967-8

RTECSNumber : WH8882000

TSCA : No

Recommended Storage : Store at -20°C

Documents