missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Triphenyltin hydride, 97%

Manufacturer:  Alfa Aesar™ L0016306

 View more versions of this product

Catalog No. AAL0016306



Description

Description

Applications
Triphenyltin hydride is used as a source of ″H·″ to generate radicals or cleave carbon-oxygen bonds. And it is also used in anti-fouling paints.

Solubility
Insoluble in water.

Notes
Air & Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

Triphenyltin hydride
26°C to 28°C
5g
C18H16Sn
MFCD00003004
3544353
351.02
162°C to 168°C (0.5mmHg)
892-20-6
>110°C (230°F)
(C6H5)3SnH
UN3146
Triphenylstannane
97%

Chemical Identifiers

892-20-6
C18H16Sn
MFCD00003004
Triphenylstannane
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.

GHS P Statement
P280h-P303+P361+P353-P305+P351+P338-P309-P310-P501a
Wear protective gloves/clothing.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.

RTECSNumber : WH8882000

Recommended Storage : −30° to −10°C

Documents