Please login to your online account to display your discounted pricing

Alfa Aesar™ Triphenyltin acetate, 97%

Chemical Identifiers

Chemical Name or Material Triphenyltin acetate
CAS 900-95-8
Molecular Formula C20H18O2Sn
Linear Formula (C6H5)3SnOOCCH3
Formula Weight 409.05g/mol
MDL Number MFCD00014977
UN Number UN3146
Beilstein 4146107
Merck Index 14,9745
Synonym Fentin acetate; Triphenylacetoxytin
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4578909

 
alfa aesar™
45789-09
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4578918

 
alfa aesar™
45789-18
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 121° to 123°C
Quantity 10g
Color White
Chemical Name or Material Triphenyltin acetate
CAS 900-95-8
Physical Form Powder
Assay 97%
Molecular Formula C20H18O2Sn
Linear Formula (C6H5)3SnOOCCH3
Formula Weight 409.05g/mol
MDL Number MFCD00014977
UN Number UN3146
Beilstein 4146107
Merck Index 14,9745
Synonym Fentin acetate; Triphenylacetoxytin
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H330-H372-H351-H361-H318-H315-H335
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
Suspected of damaging fertility or the unborn child.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P260-P301+P310-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
RTECS Number WH6650000
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H330-H372-H351-H361-H318-H315-H335
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
Suspected of damaging fertility or the unborn child.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.