missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Triphenylsulfonium Tetrafluoroborate 98.0+%, TCI America™
$185.81 - $554.11
Chemical Identifiers
CAS | 437-13-8 |
---|---|
Molecular Formula | C18H15BF4S |
Molecular Weight (g/mol) | 350.18 |
MDL Number | MFCD00671581 |
InChI Key | RTWMEMGVYYTCOZ-UHFFFAOYSA-N |
PubChem CID | 2760877 |
IUPAC Name | tetrafluoroboranuide; triphenylsulfanium |
SMILES | F[B-](F)(F)F.C1=CC=C(C=C1)[S+](C1=CC=CC=C1)C1=CC=CC=C1 |
Chemical Identifiers
437-13-8 | |
350.18 | |
RTWMEMGVYYTCOZ-UHFFFAOYSA-N | |
tetrafluoroboranuide; triphenylsulfanium |
C18H15BF4S | |
MFCD00671581 | |
2760877 | |
F[B-](F)(F)F.C1=CC=C(C=C1)[S+](C1=CC=CC=C1)C1=CC=CC=C1 |
Specifications
437-13-8 | |
White | |
MFCD00671581 | |
1759 | |
F[B-](F)(F)F.C1=CC=C(C=C1)[S+](C1=CC=CC=C1)C1=CC=CC=C1 | |
350.18 | |
350.18 | |
Crystalline Powder |
194°C | |
C18H15BF4S | |
1 g | |
RTWMEMGVYYTCOZ-UHFFFAOYSA-N | |
tetrafluoroboranuide; triphenylsulfanium | |
2760877 | |
≥98.0% (HPLC) | |
Triphenylsulfonium Tetrafluoroborate |
Safety and Handling
TSCA : No