Please login to your online account to display your discounted pricing

Alfa Aesar™ Triphenylsulfonium hexafluoroarsenate, As 14.5% min

Chemical Identifiers

Chemical Name or Material Triphenylsulfonium hexafluoroarsenate
CAS 57900-42-2
Linear Formula (C6H5)3SAsF6
Formula Weight 452.29
MDL Number MFCD00061401
UN Number UN1557
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA2084703

  SDS
alfa aesar™
2084703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA2084706

  SDS
alfa aesar™
2084706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 452.29
Color White
Health Hazard 2 GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
Health Hazard 3 GHS P Statement
P261-P301+P310-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
UN Number UN1557
Linear Formula (C6H5)3SAsF6
Sensitivity Moisture Sensitive
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.
CAS 57900-42-2
Melting Point 195° to 197°C
Physical Form Powder
Health Hazard 1 Danger
MDL Number MFCD00061401
Concentration As 14.5% min.
Quantity 1g
Chemical Name or Material Triphenylsulfonium hexafluoroarsenate
Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.