Please login to your online account to display your discounted pricing

Alfa Aesar™ Triphenylsulfonium chloride, 94%

$141.76 - $141.76

Chemical Identifiers

Chemical Name or Material Triphenylsulfonium chloride
CAS 4270-70-6
Molecular Formula (C6H5)3SCl
Formula Weight 298.84
MDL Number MFCD00031716
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2605903

 
alfa aesar™
H2605903
1g Each for $141.76
Description & Specifications

Specifications

Formula Weight 298.84
Chemical Name or Material Triphenylsulfonium chloride
Assay 94%
Molecular Formula (C6H5)3SCl
MDL Number MFCD00031716
CAS 4270-70-6
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
Triphenylsulfonium chloride is used as a key intermediate that is widely used in the new electronic chemicals such as semiconductor, LCD and TFT.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.