missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triphenylsulfonium Bromide, TCI America™

Manufacturer:  TCI America T16095G

 View more versions of this product

Catalog No. T16095G



Specifications

Specifications

Triphenylsulfonium Bromide
293°C
3353-89-7
MFCD00428762
C1=CC=C(C=C1)[S+](C2=CC=CC=C2)C3=CC=CC=C3.[Br-]
343.282
343.28
≥98.0% (T)
White
5g
C18H15BrS
VMJFYMAHEGJHFH-UHFFFAOYSA-M
triphenylsulfanium;bromide
11984010
Crystal-Powder at 20°C

Chemical Identifiers

3353-89-7
343.282
VMJFYMAHEGJHFH-UHFFFAOYSA-M
triphenylsulfanium;bromide
C18H15BrS
MFCD00428762
11984010
C1=CC=C(C=C1)[S+](C2=CC=CC=C2)C3=CC=CC=C3.[Br-]
Safety and Handling

Safety and Handling

TSCA : No

Documents