missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(Triphenylphosphoranylidene)acetaldehyde, TCI America™

Manufacturer:  TCI America T20015G

 View more versions of this product

Catalog No. T20015G



Specifications

Specifications

(Triphenylphosphoranylidene)acetaldehyde
198°C
2136-75-6
MFCD00006994
CQCAYWAIRTVXIY-UHFFFAOYSA-N
2-(triphenyl-$l^{5}-phosphanylidene)acetaldehyde
75051
Crystal-Powder at 20°C
Undesignated
5g
C20H17OP
2-triphenylphosphoranylidene acetaldehyde, formylmethylene triphenylphosphorane, triphenylphosphoranylidene acetaldehyde, formylmethylenetriphenylphosphorane, formylmethylene triphenylphosphorane, acetaldehyde, triphenylphosphoranylidene, triphenylphosphoranylidene acetaldehyde, 2-triphenyl-??-phosphanylidene acetaldehyde, acmc-209fjg, ksc492c5l
C1=CC=C(C=C1)P(=CC=O)(C2=CC=CC=C2)C3=CC=CC=C3
304.329
304.33
≥98.0% (HPLC)

Chemical Identifiers

2136-75-6
304.329
CQCAYWAIRTVXIY-UHFFFAOYSA-N
75051
C1=CC=C(C=C1)P(=CC=O)(C2=CC=CC=C2)C3=CC=CC=C3
C20H17OP
MFCD00006994
2-triphenylphosphoranylidene acetaldehyde, formylmethylene triphenylphosphorane, triphenylphosphoranylidene acetaldehyde, formylmethylenetriphenylphosphorane, formylmethylene triphenylphosphorane, acetaldehyde, triphenylphosphoranylidene, triphenylphosphoranylidene acetaldehyde, 2-triphenyl-??-phosphanylidene acetaldehyde, acmc-209fjg, ksc492c5l
2-(triphenyl-$l^{5}-phosphanylidene)acetaldehyde
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents