Please login to your online account to display your discounted pricing

Triphenylphosphine resin, 1% DVB, 100-200 mesh, 1-1.5 mmol/g, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Triphenylphosphine resin
Name Note Crosslinked with 1% DVB, 1.2-1.5 mmol/g, 100-200 mesh
CAS 39319-11-4
MDL Number MFCD00148025
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC358330250

  SDS
thermo fisher scientific
AC358330250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC358330050

  SDS
thermo fisher scientific
358330050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 25g
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Chemical Name or Material Triphenylphosphine resin
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 39319-11-4
MDL Number MFCD00148025
Name Note Crosslinked with 1% DVB, 1.2-1.5 mmol/g, 100-200 mesh
Packaging Glass bottle
Safety and Handling

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.