Please login to your online account to display your discounted pricing

Triphenylphosphine resin, 1% DVB, 100-200 mesh, 1-1.5 mmol/g, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  358330050

 View more versions of this product

Catalog No. AC358330050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Quantity 5g
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
MDL Number MFCD00148025
Name Note Crosslinked with 1% DVB, 1.2-1.5 mmol/g, 100-200 mesh
Chemical Name or Material Triphenylphosphine resin
CAS 39319-11-4
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Triphenylphosphine resin
Name Note Crosslinked with 1% DVB, 1.2-1.5 mmol/g, 100-200 mesh
CAS 39319-11-4
MDL Number MFCD00148025
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.

SDS & Safety Documents