Please login to your online account to display your discounted pricing

Triphenylphosphine resin, 1% crosslinked with DVB, 1.0-1.5mmol/g, 100-200 mesh, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC366870250

 View more versions of this product

Catalog No. AC366870250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form 100 To 200 Mesh
Quantity 25g
CAS 39319-11-4
Chemical Name or Material Triphenylphosphine resin
Name Note 1% crosslinked with DVB, 1.2-1.5mmol/g
MDL Number MFCD00148025
Packaging Glass bottle

Specifications

Chemical Name or Material Triphenylphosphine resin
Name Note 1% crosslinked with DVB, 1.2-1.5mmol/g
CAS 39319-11-4
Physical Form 100 To 200 Mesh
MDL Number MFCD00148025