Please login to your online account to display your discounted pricing

Alfa Aesar™ Triphenylphosphine, polymer-supported, 1.4-2.0 mmol/g on polystyrene

$531.30 - $531.30

Chemical Identifiers

Chemical Name or Material Triphenylphosphine
Name Note polymer-supported, 1.4 to 2.0mmol/g on polystyrene
MDL Number MFCD00148025
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1947803

 
alfa aesar™
L19478-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1947806

 
alfa aesar™
L19478-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1947814

 
alfa aesar™
L19478-14
25g Each for $531.30
Description & Specifications

Specifications

Quantity 1g
Chemical Name or Material Triphenylphosphine
Name Note polymer-supported, 1.4 to 2.0mmol/g on polystyrene
MDL Number MFCD00148025
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P363-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P363-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.