Please login to your online account to display your discounted pricing

Alfa Aesar™ Triphenylphosphine oxide, polymer-supported, 1.2-1.8 mmol/g on polystyrene

$79.72 - $79.72

Chemical Identifiers

Chemical Name or Material Triphenylphosphine oxide
Name Note polymer-supported, 1.2 to 1.8mmol/g on polystyrene
MDL Number MFCD03458802
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1947403

  SDS
alfa aesar™
L1947403
1g Each for $79.72

AAL1947406

  SDS
alfa aesar™
L1947406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1947409

  SDS
alfa aesar™
L1947409
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Triphenylphosphine oxide
Name Note polymer-supported, 1.2 to 1.8mmol/g on polystyrene
MDL Number MFCD03458802
Quantity 1g