Please login to your online account to display your discounted pricing

triphenylphosphine oxide, Maybridge


Quantity 100g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  BTB08974FA

 View more versions of this product

Catalog No. BTB08974FA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 278.29
Quantity 100g
Assay 97%
Packaging Amber glass bottle
CAS 791-28-6
Chemical Name or Material Triphenylphosphine oxide
Linear Formula (C6H5)3P(O)
Assay Percent Range 97%
Molecular Formula C18H15OP

Specifications

Chemical Name or Material Triphenylphosphine oxide
Assay 97%
Assay Percent Range 97%
Molecular Formula C18H15OP
Linear Formula (C6H5)3P(O)
Formula Weight 278.29