Please login to your online account to display your discounted pricing

triphenylphosphine oxide, Maybridge

Chemical Identifiers

Chemical Name or Material Triphenylphosphine oxide
Assay 97%
Assay Percent Range 97%
Molecular Formula C18H15OP
Linear Formula (C6H5)3P(O)
Formula Weight 278.29
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

BTB08974FA

 
fisher scientific company
BTB08974FA
Amber glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 278.29
Quantity 100g
Assay 97%
Packaging Amber glass bottle
CAS 791-28-6
Chemical Name or Material Triphenylphosphine oxide
Linear Formula (C6H5)3P(O)
Assay Percent Range 97%
Molecular Formula C18H15OP