missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(Triphenylphosphine)gold(I) Chloride, TCI America™

Manufacturer:  TCI America T29941G

 View more versions of this product

Catalog No. T29941G



Specifications

Specifications

(Triphenylphosphine)gold(I) Chloride
247°C
14243-64-2
MFCD00009588
IFPWCRBNZXUWGC-UHFFFAOYSA-M
chlorogold;triphenylphosphane
10874691
Crystal-Powder at 20°C
White-Yellow
1g
C18H15AuClP
chloro triphenylphosphine gold i, chloro triphenylphosphine gold, chlorotriphenylphosphine gold i, triphenylphosphinegold i chloride, aucl pph3, pubchem16017, ph3p aucl, chlorogold; triphenylphosphane, chlorogold; triphenylphosphine, chloranylgold; triphenylphosphane
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.Cl[Au]
494.708
494.71
≥95.0% (T)

Chemical Identifiers

14243-64-2
494.708
IFPWCRBNZXUWGC-UHFFFAOYSA-M
10874691
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.Cl[Au]
C18H15AuClP
MFCD00009588
chloro triphenylphosphine gold i, chloro triphenylphosphine gold, chlorotriphenylphosphine gold i, triphenylphosphinegold i chloride, aucl pph3, pubchem16017, ph3p aucl, chlorogold; triphenylphosphane, chlorogold; triphenylphosphine, chloranylgold; triphenylphosphane
chlorogold;triphenylphosphane
Safety and Handling

Safety and Handling

TSCA : No

Documents