missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Triphenylphosphine diiodide, tech. 90%

$45.20 - $652.62

Chemical Identifiers

CAS 6396-07-2
Molecular Formula C18H15I2P
Molecular Weight (g/mol) 516.10
MDL Number MFCD00216568
InChI Key NSWZEXJQHIXCFL-UHFFFAOYSA-N
Synonym 4-16-00-01013 beilstein handbook reference
PubChem CID 201622
IUPAC Name diiodo(triphenyl)-$l^{5}-phosphane
SMILES C1=CC=C(C=C1)P(C2=CC=CC=C2)(C3=CC=CC=C3)(I)I
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5558503 View Documents Alfa Aesar™
H5558503
1g Each for $45.20
 
AAH5558514 View Documents Alfa Aesar™
H5558514
25g Each for $652.62
 
Specifications

Chemical Identifiers

6396-07-2
516.10
NSWZEXJQHIXCFL-UHFFFAOYSA-N
201622
C1=CC=C(C=C1)P(C2=CC=CC=C2)(C3=CC=CC=C3)(I)I
C18H15I2P
MFCD00216568
4-16-00-01013 beilstein handbook reference
diiodo(triphenyl)-$l^{5}-phosphane

Specifications

Light Sensitive
210°C to 211°C
6396-07-2
(C6H5)3PI2
UN2923
4-16-00-01013 beilstein handbook reference
C1=CC=C(C=C1)P(C2=CC=CC=C2)(C3=CC=CC=C3)(I)I
516.10
516.1
90%
Triphenylphosphine diiodide
Yellow to Orange
1g
C18H15I2P
MFCD00216568
2740417
NSWZEXJQHIXCFL-UHFFFAOYSA-N
diiodo(triphenyl)-$l^{5}-phosphane
201622
Powder
Technical
Safety and Handling

Safety and Handling

GHS H Statement
H334-H361-H314-H318-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of damaging fertility or the unborn child.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.

GHS P Statement
P260-P285-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
In case of inadequate ventilation wear respiratory protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

RTECSNumber : TB6100000