missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Triphenylphosphine diiodide, tech. 90%

Manufacturer:  Alfa Aesar™ H5558514

 View more versions of this product

Catalog No. AAH5558514



Specifications

Specifications

Triphenylphosphine diiodide
Yellow to Orange
25g
C18H15I2P
MFCD00216568
2740417
NSWZEXJQHIXCFL-UHFFFAOYSA-N
diiodo(triphenyl)-$l^{5}-phosphane
201622
Powder
Technical
Light Sensitive
210°C to 211°C
6396-07-2
(C6H5)3PI2
UN2923
4-16-00-01013 beilstein handbook reference
C1=CC=C(C=C1)P(C2=CC=CC=C2)(C3=CC=CC=C3)(I)I
516.10
516.1
90%

Chemical Identifiers

6396-07-2
516.10
NSWZEXJQHIXCFL-UHFFFAOYSA-N
201622
C1=CC=C(C=C1)P(C2=CC=CC=C2)(C3=CC=CC=C3)(I)I
C18H15I2P
MFCD00216568
4-16-00-01013 beilstein handbook reference
diiodo(triphenyl)-$l^{5}-phosphane
Safety and Handling

Safety and Handling

GHS H Statement
H334-H361-H314-H318-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of damaging fertility or the unborn child.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.

GHS P Statement
P260-P285-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
In case of inadequate ventilation wear respiratory protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

RTECSNumber : TB6100000

Documents