Please login to your online account to display your discounted pricing

Alfa Aesar™ Triphenylphosphine dichloride, 96%

$64.93 - $64.93

Chemical Identifiers

Chemical Name or Material Triphenylphosphine dichloride
CAS 2526-64-9
Molecular Formula (C6H5)3PCl2
Formula Weight 333.19
MDL Number MFCD00010359
UN Number UN3261
Synonym Dichlorotriphenylphosphorane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2743622

  SDS
alfa aesar™
H2743622
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2743614

  SDS
alfa aesar™
H2743614*
25g Each for $64.93
Description & Specifications

Specifications

Formula Weight 333.19
Melting Point 84° to 86°C
UN Number UN3261
Molecular Formula (C6H5)3PCl2
Assay 96%
Quantity 100g
MDL Number MFCD00010359
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Danger
Synonym Dichlorotriphenylphosphorane
Chemical Name or Material Triphenylphosphine dichloride
CAS 2526-64-9

Applications
As a precursor to fire-retardant materials. Mainly used as a chlorinating agent widely used in organic chemistry. Applications include the conversion of alcohols and ethers to alkyl chlorides, the cleavage of epoxides to vicinal dichlorides and the chlorination of carboxylic acids to acyl chlorides.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases, air, moisture.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.