missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triphenylphosphine dibromide, ca. 33% bromine, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC291071000

 View more versions of this product

Catalog No. AC291071000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

C-X bond formation reagent
Specifications

Specifications

Triphenylphosphine dibromide
Plastic bottle
235.0°C
ca. 33% bromine
C18H15Br2P
MFCD00000054
01,1247; 13,333
bromotriphenylphosphonium bromide, dibromo triphenylphosphorane, dibromotriphenyl-??-phosphane, dibromotriphenylphosphine, dibromotriphenylphosphorane, dibromtriphenylphosphorane, phosphorane, dibromotriphenyl, triphenyldibromophosphorane, triphenylphosphinedibromide, unii-7y0j6r63jd
C1=CC=C(C=C1)P(C2=CC=CC=C2)(C3=CC=CC=C3)(Br)Br
422.09
422.09
ca. 33% bromine
Authentic
Pink to Yellow
100g
1034-39-5
(C6H5)3PBr2
16, 864
15, 9917
OCXGTPDKNBIOTF-UHFFFAOYSA-N
dibromo(triphenyl)-$l^{5}-phosphane
70579
Crystalline Powder

Chemical Identifiers

1034-39-5
422.09
OCXGTPDKNBIOTF-UHFFFAOYSA-N
70579
C1=CC=C(C=C1)P(C2=CC=CC=C2)(C3=CC=CC=C3)(Br)Br
C18H15Br2P
MFCD00000054
bromotriphenylphosphonium bromide, dibromo triphenylphosphorane, dibromotriphenyl-??-phosphane, dibromotriphenylphosphine, dibromotriphenylphosphorane, dibromtriphenylphosphorane, phosphorane, dibromotriphenyl, triphenyldibromophosphorane, triphenylphosphinedibromide, unii-7y0j6r63jd
dibromo(triphenyl)-$l^{5}-phosphane
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 213-855-1

SDS Documents