missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triphenylphosphine dibromide, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1155209

 View more versions of this product

Catalog No. AAA1155209



Description

Description

Triphenylphosphine dibromide is used as a reagent for conversion of aldoximes into nitriles. It acts as a reagent for cleavage of acetals. It reacts with amines to give iminophosphoranes. Further, it serves as a reagent in the preparation of alkyl and aryl bromides from alcohols and phenols without rearrangement.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenylphosphine dibromide is used as a reagent for conversion of aldoximes into nitriles. It acts as a reagent for cleavage of acetals. It reacts with amines to give iminophosphoranes. Further, it serves as a reagent in the preparation of alkyl and aryl bromides from alcohols and phenols without rearrangement.

Solubility
Soluble in water, dichloromethane, acetonitrile, phenyl cyanide, dimethyl formamide, ether, chloroform, alcohol, carbon tetrachloride, acetic acid and carbon disulfide. Sparingly soluble in phenyl chloride, triphenylphosphine and phenyl hydride. Insoluble in aqueous bromine.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong bases, strong oxidizing agents, bases and active metals.
Specifications

Specifications

Triphenylphosphine dibromide
∼260°C (decomposition)
MFCD00000054
UN3261
Moisture sensitive
Soluble in water,dichloromethane,acetonitrile,phenyl cyanide,dimethyl formamide,ether,chloroform,alcohol,carbon tetrachloride,acetic acid and carbon disulfide. Sparingly soluble in phenyl chloride,triphenylphosphine and phenyl hydride. Insoluble in aqueous bromine.
BrP(Br)(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
422.10
422.1
1034-39-5
C18H15Br2P
10 g
917652
dibromotriphenylphosphorane, phosphorane, dibromotriphenyl, triphenyldibromophosphorane, unii-7y0j6r63jd, bromotriphenylphosphonium bromide, dibromotriphenyl-??-phosphane, dibromotriphenylphosphine, dibromtriphenylphosphorane, triphenylphosphinedibromide, dibromo triphenylphosphorane
OCXGTPDKNBIOTF-UHFFFAOYSA-N
dibromo(triphenyl)-$l^{5}-phosphane
70579
96%

Chemical Identifiers

1034-39-5
422.10
OCXGTPDKNBIOTF-UHFFFAOYSA-N
70579
BrP(Br)(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
C18H15Br2P
MFCD00000054
dibromotriphenylphosphorane, phosphorane, dibromotriphenyl, triphenyldibromophosphorane, unii-7y0j6r63jd, bromotriphenylphosphonium bromide, dibromotriphenyl-??-phosphane, dibromotriphenylphosphine, dibromtriphenylphosphorane, triphenylphosphinedibromide, dibromo triphenylphosphorane
dibromo(triphenyl)-$l^{5}-phosphane
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P370+P380-P501c

H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 213-855-1

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only