missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Triphenylmethyl mercaptan, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 140415000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Triphenylmethyl mercaptan | |
97.5% min. | |
96% min. (HPLC) | |
C19H16S | |
MFCD00004854 | |
06, I, 352 | |
Solubility in water: insoluble. Other solubilities: soluble in toluene | |
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)S | |
276.39 | |
276.39 |
3695-77-0 | |
Authentic | |
Glass bottle | |
(C6H5)3CSH | |
500g | |
triphenylmethyl mercaptan, trityl mercaptan, tritylthiol, triphenylmethylmercaptan, methanethiol, triphenyl, tritylmercaptan, triphenyl-methanethiol, alpha,alpha-diphenylbenzenemethanethiol, benzenemethanethiol, alpha,alpha-diphenyl, benzenemethanethiol, .alpha.,.alpha.-diphenyl | |
JQZIKLPHXXBMCA-UHFFFAOYSA-N | |
triphenylmethanethiol | |
77281 | |
97% |
Chemical Identifiers
3695-77-0 | |
276.39 | |
JQZIKLPHXXBMCA-UHFFFAOYSA-N | |
77281 | |
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)S |
C19H16S | |
MFCD00004854 | |
triphenylmethyl mercaptan, trityl mercaptan, tritylthiol, triphenylmethylmercaptan, methanethiol, triphenyl, tritylmercaptan, triphenyl-methanethiol, alpha,alpha-diphenylbenzenemethanethiol, benzenemethanethiol, alpha,alpha-diphenyl, benzenemethanethiol, .alpha.,.alpha.-diphenyl | |
triphenylmethanethiol |
Safety and Handling
GHS H Statement
Harmful if inhaled.
GHS Signal Word: Warning
EINECSNumber : 223-020-3
RUO â Research Use Only