Please login to your online account to display your discounted pricing

Triphenylmethylemercaptan, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  140410250

 View more versions of this product

Catalog No. AC140410250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 276.39
Physical Form Powder
Quantity 25g
Color Cream to pale yellow
Beilstein 06, I, 352
Melting Point 103° to 107°C
Linear Formula (C6H5)3CSH
MDL Number MFCD00004854
Molecular Formula C19H16S
Assay Percent Range 98%
CAS 3695-77-0
Chemical Name or Material Triphenylmethylemercaptan
Synonym Triphenylmethanethiol
Infrared Spectrum Authentic
Elemental Analysis 97.5% min.
Packaging Glass bottle

Specifications

Chemical Name or Material Triphenylmethylemercaptan
Assay Percent Range 98%
Molecular Formula C19H16S
Linear Formula (C6H5)3CSH
Formula Weight 276.39
MDL Number MFCD00004854
Synonym Triphenylmethanethiol