Please login to your online account to display your discounted pricing

Triphenylmethylebromide, 98%, ACROS Organics™


Quantity 250g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC140392500

 View more versions of this product

Catalog No. AC140392500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Formula Weight 323.22
Assay ≥97.5%
Quantity 250g
Color Light yellow
Melting Point 151° to 155°C
Beilstein 05, 704
Fieser 01,1254; 16,375
Packaging Glass bottle
Infrared Spectrum Authentic
Synonym Bromotriphenylmethane
Linear Formula (C6H5)3CBr
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
MDL Number MFCD00000120
Assay Percent Range 98%
Chemical Name or Material Triphenylmethylebromide
CAS 596-43-0
Molecular Formula C19H15Br
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Triphenylmethylebromide
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C19H15Br
Linear Formula (C6H5)3CBr
Formula Weight 323.22
MDL Number MFCD00000120
Synonym Bromotriphenylmethane
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents