missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triphenylmethyl bromide, 98%, Thermo Scientific™

$58.66 - $212.59

Chemical Identifiers

CAS 596-43-0
Molecular Formula C19H15Br
Molecular Weight (g/mol) 323.22
MDL Number MFCD00000120
InChI Key NZHXEWZGTQSYJM-UHFFFAOYSA-N
Synonym 2-bromo-1,1,1-triphenylethane, benzene, 1,1',1-bromomethylidyne tris, bromodiphenylmethyl benzene, bromomethanetriyl tribenzene, bromotriphenylmethane, methane, bromotriphenyl, triphenylbromomethane, triphenylmethyl bromide, triphenylmethylbromide, trityl bromide
PubChem CID 11692
IUPAC Name [bromo(diphenyl)methyl]benzene
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC140390100
SDS View Documents Promotion Details
Thermo Fisher Scientific
140390100
10g Glass bottle
Each for $58.66
 
AC140390500
SDS View Documents Promotion Details
Thermo Fisher Scientific
140390500
50g Glass bottle
Each for $212.59
 
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

596-43-0
323.22
NZHXEWZGTQSYJM-UHFFFAOYSA-N
11692
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)Br
C19H15Br
MFCD00000120
2-bromo-1,1,1-triphenylethane, benzene, 1,1',1-bromomethylidyne tris, bromodiphenylmethyl benzene, bromomethanetriyl tribenzene, bromotriphenylmethane, methane, bromotriphenyl, triphenylbromomethane, triphenylmethyl bromide, triphenylmethylbromide, trityl bromide
[bromo(diphenyl)methyl]benzene

Specifications

596-43-0
151°C to 155°C
97.5% min. (Argentometry)
C19H15Br
MFCD00000120
Glass bottle
2-bromo-1,1,1-triphenylethane, benzene, 1,1',1-bromomethylidyne tris, bromodiphenylmethyl benzene, bromomethanetriyl tribenzene, bromotriphenylmethane, methane, bromotriphenyl, triphenylbromomethane, triphenylmethyl bromide, triphenylmethylbromide, trityl bromide
Solubility in water: insoluble
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)Br
323.22
323.22
98%
Yellow
230°C (15.0mmHg)
Authentic
(C6H5)3CBr
05, 704
01,1254; 16,375
10g
NZHXEWZGTQSYJM-UHFFFAOYSA-N
[bromo(diphenyl)methyl]benzene
11692
Crystalline Powder
Triphenylmethyl bromide
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 209-884-4

SDS
Product Certifications

RUO – Research Use Only