Please login to your online account to display your discounted pricing

Triphenylmethylebromide, 98%, ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  140390500

 View more versions of this product

Catalog No. AC140390500

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 323.22
Quantity 50g
Color Light yellow
Beilstein 05, 704
Fieser 01,1254; 16,375
Melting Point 151° to 155°C
Packaging Glass bottle
CAS 596-43-0
Synonym Bromotriphenylmethane
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Triphenylmethylebromide
Assay Percent Range 98%
Molecular Formula C19H15Br
Linear Formula (C6H5)3CBr
MDL Number MFCD00000120
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Triphenylmethylebromide
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C19H15Br
Linear Formula (C6H5)3CBr
Formula Weight 323.22
MDL Number MFCD00000120
Synonym Bromotriphenylmethane
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents