Please login to your online account to display your discounted pricing

Alfa Aesar™ (Triphenylmethyl)triphenylphosphonium chloride, 98%

$78.04 - $1,069.80

Chemical Identifiers

Chemical Name or Material (Triphenylmethyl)triphenylphosphonium chloride
CAS 48234-64-6
Molecular Formula (C6H5)3CP(C6H5)3Cl
Formula Weight 541.07
MDL Number MFCD00082244
Beilstein 389941
Synonym Trityl triphenylphosphonium chloride; Trityltriphenylphosphonium chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2231909

  SDS
alfa aesar™
B2231909
10g Each for $78.04

AAB2231918

  SDS
alfa aesar™
B2231918
50g Each for $278.61

AAB2231930

  SDS
alfa aesar™
B2231930
250g Each for $1,069.80
Description & Specifications

Specifications

Formula Weight 541.07
Chemical Name or Material (Triphenylmethyl)triphenylphosphonium chloride
Assay 98%
Molecular Formula (C6H5)3CP(C6H5)3Cl
MDL Number MFCD00082244
CAS 48234-64-6
Beilstein 389941
Synonym Trityl triphenylphosphonium chloride; Trityltriphenylphosphonium chloride
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 244° to 245°C
Quantity 10g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.