Learn More
Triphenylmethyl mercaptan, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1488822
Description
Triphenylmethyl mercaptan, is used as a reagent used for the introduction of thiol group. Triphenylmethanethiol is a reagent used for the mild introduction of the mercapto functionality. It is also used as a pharmaceutical intermediate; Chemical synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTriphenylmethyl mercaptan, is used as a reagent used for the introduction of thiol group. Triphenylmethanethiol is a reagent used for the mild introduction of the mercapto functionality. It is also used as a pharmaceutical intermediate; Chemical synthesis.
Solubility
It is insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under recommended storage conditions.
Specifications
Triphenylmethyl mercaptan | |
103°C to 108°C | |
C19H16S | |
100 g | |
triphenylmethyl mercaptan, trityl mercaptan, tritylthiol, triphenylmethylmercaptan, methanethiol, triphenyl, tritylmercaptan, triphenyl-methanethiol, alpha,alpha-diphenylbenzenemethanethiol, benzenemethanethiol, alpha,alpha-diphenyl, benzenemethanethiol, .alpha.,.alpha.-diphenyl | |
JQZIKLPHXXBMCA-UHFFFAOYSA-N | |
triphenylmethanethiol | |
77281 | |
≥98% |
3695-77-0 | |
Stench | |
MFCD00004854 | |
2113168 | |
It is insoluble in water. | |
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)S | |
276.397 | |
276.4 |
Chemical Identifiers
3695-77-0 | |
276.397 | |
JQZIKLPHXXBMCA-UHFFFAOYSA-N | |
77281 | |
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)S |
C19H16S | |
MFCD00004854 | |
triphenylmethyl mercaptan, trityl mercaptan, tritylthiol, triphenylmethylmercaptan, methanethiol, triphenyl, tritylmercaptan, triphenyl-methanethiol, alpha,alpha-diphenylbenzenemethanethiol, benzenemethanethiol, alpha,alpha-diphenyl, benzenemethanethiol, .alpha.,.alpha.-diphenyl | |
triphenylmethanethiol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P271-P304+P340-P312
H332
EINECSNumber : 223-020-3
RTECSNumber : PB5335000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only