Please login to your online account to display your discounted pricing

Triphenylmethyl Chloride 98%, ACROS Organics™

$56.31 - $171.62

Chemical Identifiers

Chemical Name or Material Triphenylmethyl chloride
Name Note 98%
CAS 76-83-5
Assay ≥97.5% (ex Cl)
Assay Percent Range 98%
Molecular Formula C19H15Cl
Linear Formula (C6H5)3CCl
Formula Weight 278.77
MDL Number MFCD00000813
Synonym Triphenylchloromethane
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC140400025

  SDS
thermo fisher scientific
AC140400025
Plastic bottle 2.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC140401000

  SDS
thermo fisher scientific
140401000
Plastic bottle 100g Each for $56.31

AC140405000

  SDS
thermo fisher scientific
140405000
Plastic bottle 500g Each for $171.62
Description & Specifications

Specifications

Assay ≥97.5% (ex Cl)
Physical Form Powder or Crystals
Formula Weight 278.77
Quantity 2.5kg
Color White to off-white or light yellow-beige
TSCA TSCA
EINECS Number 200-986-4
RTECS Number PA6450000
Melting Point 109° to 113°C
Beilstein 05, 700
Merck Index 15, 9944
Fieser 01,1254; 02,453; 04,565; 05,741; 06,656; 15,359
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Plastic bottle
Synonym Triphenylchloromethane
CAS 76-83-5
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Very toxic to aquatic life.
Assay Percent Range 98%
Name Note 98%
MDL Number MFCD00000813
Chemical Name or Material Triphenylmethyl chloride
Linear Formula (C6H5)3CCl
Molecular Formula C19H15Cl
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Very toxic to aquatic life.