Please login to your online account to display your discounted pricing

Triphenylmethyl Chloride 98%, ACROS Organics™


Quantity 500g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  140405000

 View more versions of this product

Catalog No. AC140405000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5% (ex Cl)
Physical Form Powder or Crystals
Formula Weight 278.77
Quantity 500g
Color White to off-white or light yellow-beige
Beilstein 05, 700
Merck Index 15, 9944
EINECS Number 200-986-4
TSCA TSCA
RTECS Number PA6450000
Melting Point 109° to 113°C
Fieser 01,1254; 02,453; 04,565; 05,741; 06,656; 15,359
Chemical Name or Material Triphenylmethyl chloride
Packaging Plastic bottle
Name Note 98%
Assay Percent Range 98%
Molecular Formula C19H15Cl
Linear Formula (C6H5)3CCl
MDL Number MFCD00000813
Synonym Triphenylchloromethane
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Very toxic to aquatic life.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
CAS 76-83-5

Specifications

Chemical Name or Material Triphenylmethyl chloride
Name Note 98%
CAS 76-83-5
Assay ≥97.5% (ex Cl)
Assay Percent Range 98%
Molecular Formula C19H15Cl
Linear Formula (C6H5)3CCl
Formula Weight 278.77
MDL Number MFCD00000813
Synonym Triphenylchloromethane
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Very toxic to aquatic life.

SDS & Safety Documents