missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Triphenylmethanethiol, TCI America™

Manufacturer:  TCI America T224425G

 View more versions of this product

Catalog No. T224425G



Specifications

Specifications

Triphenylmethanethiol
108°C
3695-77-0
MFCD00004854
JQZIKLPHXXBMCA-UHFFFAOYSA-N
triphenylmethanethiol
77281
Crystal-Powder at 20°C
Yellow
25g
C19H16S
triphenylmethyl mercaptan, trityl mercaptan, tritylthiol, triphenylmethylmercaptan, methanethiol, triphenyl, tritylmercaptan, triphenyl-methanethiol, alpha,alpha-diphenylbenzenemethanethiol, benzenemethanethiol, alpha,alpha-diphenyl, benzenemethanethiol, .alpha.,.alpha.-diphenyl
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)S
276.397
276.40
≥97.0% (HPLC)

Chemical Identifiers

3695-77-0
276.397
JQZIKLPHXXBMCA-UHFFFAOYSA-N
77281
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)S
C19H16S
MFCD00004854
triphenylmethyl mercaptan, trityl mercaptan, tritylthiol, triphenylmethylmercaptan, methanethiol, triphenyl, tritylmercaptan, triphenyl-methanethiol, alpha,alpha-diphenylbenzenemethanethiol, benzenemethanethiol, alpha,alpha-diphenyl, benzenemethanethiol, .alpha.,.alpha.-diphenyl
triphenylmethanethiol
Safety and Handling

Safety and Handling

RTECSNumber : PB5335000

TSCA : Yes

Documents